ContactUsLinkComponent
Roll and Grow* Plating Beads 2 tubes
Roll & Grow™ Spherical Glass Plating Beads, MP Biomedicals
# de catalogue CA76045-838
Fournisseur:  MP Biomedicals
Roll and Grow* Plating Beads 2 tubes
Roll & Grow™ Spherical Glass Plating Beads, MP Biomedicals
# de catalogue CA76045-838
Fournisseur:  MP Biomedicals
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Spherical Glass Plating Beads
  • Packaging:
    Two 50 ml tubes
Spécifications
A propos de cet article
Roll & Grow™ Plating Beads are used to spread 25–400μL of bacterial or yeast cells across the surface of an agar plate.
  • Sterile
  • Easy to use
  • Spread cells in just seconds per plate
  • Provides even colony distribution
  • Eliminates hazards of flame and ethanol
Roll & Grow™ Plating Beads provide a safer, faster alternative to the traditional method of plating with a glass rod sterilized with ethanol and flame. Sterile Roll & Grow® Plating Beads are provided in two skirted 50mL conical tubes. Between six and seven 4mm glass beads are used per plate for a total of approximately 160 standard 100mm plates.
Customers Who Bought This Also Bought