Graduated Reagent Bottle
# de catalogue 470228-990
Fournisseur: Wards
Spécifications
- Capacity:1000 ml
- Color:Amber
- Cat. no.:470228-990
Spécifications
A propos de cet article
Mess-free pouring.
- Screw cap
- Borosilicate glass
- Poring ring
- Both cap and bottle are autoclavable
These economy graduated reagent bottles are the perfect addition to any laboratory. Made from borosilicate glass, these bottles have a convienent screw cap and a drip-free pouring ring to help maintain a clean work environment.
Customers Who Bought This Also Bought