ContactUsLinkComponent
Reagent Bottle Amber 1000 ml
Graduated Reagent Bottle
# de catalogue 470228-990
Fournisseur:  Wards
Reagent Bottle Amber 1000 ml
Graduated Reagent Bottle
# de catalogue 470228-990
Fournisseur:  Wards
Spécifications
  • Capacity:
    1000 ml
  • Color:
    Amber
  • Cat. no.:
    470228-990
Spécifications
A propos de cet article
Mess-free pouring.
  • Screw cap
  • Borosilicate glass
  • Poring ring
  • Both cap and bottle are autoclavable
These economy graduated reagent bottles are the perfect addition to any laboratory. Made from borosilicate glass, these bottles have a convienent screw cap and a drip-free pouring ring to help maintain a clean work environment.
Customers Who Bought This Also Bought