Order Entry
ContactUsLinkComponent
DNA Elution Buffer, 4ml
DNA Elution Buffer, Zymo Research
Catalog # 76020-524
Supplier:  Zymo Research
CAS Number:  
DNA Elution Buffer, 4ml
DNA Elution Buffer, Zymo Research
Catalog # 76020-524
Supplier:  Zymo Research
CAS Number:  

Specifications

  • Size:
    4 mL
  • Brand name:
    Zymo Research
  • Cat. no.:
    76020-524

Specifications

About this item

Customers Who Bought This Also Bought