Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
EZFlow® Syringe Filter, PES, Sterile, Foxx Life Sciences | Avantor

EZFlow® Syringe Filter, PES, Sterile, Foxx Life Sciences

25 mm, 0.22 µm, Sterile
Catalog # 76018-868
Supplier:   Foxx Life Sciences
Fournisseur #:  371-2215-OEM
EZFlow® Syringe Filter, PES, Sterile, Foxx Life Sciences
Catalog # 76018-868
Supplier:  Foxx Life Sciences
EZFlow® Syringe Filter, PES, Sterile, Foxx Life Sciences
EZFlow® Syringe Filter, PES, Sterile, Foxx Life Sciences

25 mm, 0.22 µm, Sterile

Price
$428.49
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Pack of 100
Product Details & Documents

High performance PES syringe filters have super high flow rates, very low extractables, and are very low protein binding.

  • Super high flow rates
  • Very low protein binding
  • Very low extractables
  • Perfect for use with media, buffers, and DNA

EZFlow® 13, 25, and 33 mm Syringe Filters are used for prefiltration, clarification, sterilization, or fine particulate removal. These syringe filters are available in a variety of filter membrane materials in both 0.2 and 0.45 µm pore sizes. The filter membranes are reinforced with polypropylene housing, with a female Luer-lok inlet and a male Luer slip outlet.

All at an affordable cost.

Certifications: All materials comply with USP Class VI Standards and FDA compliances.

Documents

Spécifications
  • Ø:
    25 mm
  • Taille de pore:
    0.22 µm
  • Sterility:
    Sterile
Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.