White Polypropylene Screw Caps, F217 & Fluoropolymer Resin Liner, Qorpak®
Catalogue # 76012-832
Fournisseur: Qorpak
Spécifications
- Cap Size:22-400
- Cat. no.:76012-832
Spécifications
À propos de cet article
Polypropylene caps with F217 & Fluoropolymer Resin liners are an economical alternative to Thermoset & Phenolic Fluoropolymer Resin lined caps.
- Liners are very resilient – great sealing capability
- Liners have strong resistance to chemicals and moisture
Some uses for these F-217/PTFE lined caps include analytical lab samples, high purity chemicals, strong acids solvents, environmental samples, pharmaceuticals, and diagnostic reagents.
Frequently Bought Together