ContactUsLinkComponent
White Polypropylene Screw Caps, F217 & Fluoropolymer Resin Liner, Qorpak®
Catalogue # 76012-832
Fournisseur:  Qorpak
undefined
White Polypropylene Screw Caps, F217 & Fluoropolymer Resin Liner, Qorpak®
Catalogue # 76012-832
Fournisseur:  Qorpak
Supplier Number:  272191
Spécifications
  • Cap Size:
    22-400
  • Cat. no.:
    76012-832
Spécifications
À propos de cet article
Polypropylene caps with F217 & Fluoropolymer Resin liners are an economical alternative to Thermoset & Phenolic Fluoropolymer Resin lined caps.
  • Liners are very resilient – great sealing capability
  • Liners have strong resistance to chemicals and moisture
Some uses for these F-217/PTFE lined caps include analytical lab samples, high purity chemicals, strong acids solvents, environmental samples, pharmaceuticals, and diagnostic reagents.
Frequently Bought Together