Order Entry
Canada
ContactUsLinkComponent
VWR® Polygon Spinbar Magnetic Stirring Bars
Catalogue # 76006-406
Fournisseur:  VWR International
undefined
VWR® Polygon Spinbar Magnetic Stirring Bars
Catalogue # 76006-406
Fournisseur:  VWR International

Spécifications

  • Coating:
    PTFE
  • Ø:
    8 mm (⁵/₁₆")
  • Length:
    50 mm (2")
  • Cat. no.:
    76006-406
  • Shape:
    Polygon

Spécifications

À propos de cet article

These stirbars have polygon multifaceted surfaces that add turbulence relative to similar smooth size cylindrical bars.

  • Center molded pivot ring minimizes the contact area between the bar and the vessel; reduces friction and chattering
  • Marring of plastic containers is diminished
  • Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE low friction coating; durable and inert
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclave safe
Frequently Bought Together