Spécifications
- Format:Spin column
- Description:Zymo-Spin™ IIC
- Size:50 pack
Spécifications
A propos de cet article
Spin columns for the purification of DNA or RNA.
- Format: DNA/RNA binding
- DNA binding capacity: 25 µg
- RNA binding capacity: 50 µg
- Compatibility: microcentrifuge, vacuum manifold
The Zymo-Spin™ IIC column can be used either in microcentrifuges or on vacuum manifolds for the purification of DNA and/or RNA. The Zymo-Spin™ IIC features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 25 μg of DNA, and 50 μg of RNA, in ≥ 25 μL eluate. Capacity is 900 μL.