ContactUsLinkComponent
ZR-96 MagStand 96 well
ZR-96 MagStand, Zymo Research
# de catalogue 76005-120
Fournisseur:  Zymo Research
ZR-96 MagStand, Zymo Research
# de catalogue 76005-120
Fournisseur:  Zymo Research
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    ZR-96 MagStand
  • Size:
    96 well
  • Cat. no.:
    76005-120
Spécifications
A propos de cet article
The ZR-96 MagStand is a magnetic stand for the separation of magnetic beads in standard 96-well format plates, especially for EZ-96 DNA Methylation MagPrep product line, ZR-96 Genomic DNA MagPrep, Zyppy-96 Plasmid MagBead Miniprep, and Direct-zol-96 MagBead RNA.
Customers Who Bought This Also Bought