Spécifications
- Format:Spin column/filter assembly
- Description:Zymo-Spin™ V with Reservoir
- Size:50 pack
Spécifications
A propos de cet article
Columns and filters for the purification of DNA and/or RNA from high-volume sample inputs.
- For purifying DNA and or/RNA from high-volume sample inputs.
- High-recovery.
The Zymo-Spin™ V with Reservoir assembly can be used in conjunction with centrifuges and on vacuum manifolds for the purification of DNA and/or RNA
The Zymo-Spin™ VI with Reservoir assembly can be used with vacuum manifolds for the purification of DNA and/or RNA. The spin-column and reservoir feature durable polypropylene construction. The spin-column features a unique silica-based matrix for the purification of up to 500 μg DNA or RNA in ≥ 1 mL elution buffer or water. The capacity of the spin-column with filter is 75 mL.
The Zymo-Spin™ VI with Zymo Maxi Filter™ assembly can be used with vacuum manifolds for the purification of DNA and/or RNA. The spin-column and filter feature durable polypropylene construction. The spin-column features a unique silica-based matrix for the purification of up to 500 μg DNA or RNA in ≥ 1 mL elution buffer or water. The capacity of the spin-column with filter is 75 mL.
The ZRC-GF Filter™ syringe filter features durable polypropylene construction and contains a 1.6μm pore size glass fiber filtration membrane. The filter is ideal for separating the cellular component from biological liquids (e.g., urine) and is the same filter featured in the ZR Urine DNA and RNA Isolation kits.