ContactUsLinkComponent
Zymo-Spin* V with Reservoir assembly DNA/RNA binding
Zymo-Spin™ Column/Filter Assemblies, Zymo Research
# de catalogue 76004-192
Fournisseur:  Zymo Research
Numéro CAS:  
Zymo-Spin* V with Reservoir assembly DNA/RNA binding
Zymo-Spin™ Column/Filter Assemblies, Zymo Research
# de catalogue 76004-192
Fournisseur:  Zymo Research
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Format:
    Spin column/filter assembly
  • Description:
    Zymo-Spin™ V with Reservoir
  • Size:
    50 pack
Spécifications
A propos de cet article
Columns and filters for the purification of DNA and/or RNA from high-volume sample inputs.
  • For purifying DNA and or/RNA from high-volume sample inputs.
  • High-recovery.
The Zymo-Spin™ V with Reservoir assembly can be used in conjunction with centrifuges and on vacuum manifolds for the purification of DNA and/or RNA
The Zymo-Spin™ VI with Reservoir assembly can be used with vacuum manifolds for the purification of DNA and/or RNA. The spin-column and reservoir feature durable polypropylene construction. The spin-column features a unique silica-based matrix for the purification of up to 500 μg DNA or RNA in ≥ 1 mL elution buffer or water. The capacity of the spin-column with filter is 75 mL.
The Zymo-Spin™ VI with Zymo Maxi Filter™ assembly can be used with vacuum manifolds for the purification of DNA and/or RNA. The spin-column and filter feature durable polypropylene construction. The spin-column features a unique silica-based matrix for the purification of up to 500 μg DNA or RNA in ≥ 1 mL elution buffer or water. The capacity of the spin-column with filter is 75 mL.
The ZRC-GF Filter™ syringe filter features durable polypropylene construction and contains a 1.6μm pore size glass fiber filtration membrane. The filter is ideal for separating the cellular component from biological liquids (e.g., urine) and is the same filter featured in the ZR Urine DNA and RNA Isolation kits.
Caution: For purifying DNA and or/RNA from high-volume sample inputs. High-recovery.
Customers Who Bought This Also Bought