ContactUsLinkComponent
Zymo-Spin* IC Column DNA/RNA binding
Zymo-Spin™ IC Columns, Zymo Research
# de catalogue 76004-136
Fournisseur:  Zymo Research
Numéro CAS:  
Zymo-Spin* IC Column DNA/RNA binding
Zymo-Spin™ IC Columns, Zymo Research
# de catalogue 76004-136
Fournisseur:  Zymo Research
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Format:
    Spin column
  • Description:
    Zymo-Spin™ IC
  • Size:
    50 pack
Spécifications
A propos de cet article
Spin columns for the purification of DNA or RNA.
  • Format: DNA/RNA binding
  • DNA binding capacity: 5 µg
  • RNA binding capacity: 10 µg
  • Compatibility: microcentrifuge, vacuum manifold
Capped version of the Zymo-Spin™ I column. The Zymo-Spin™ IC column can be used either in microcentrifuges or on vacuum manifolds for the purification of DNA and/or RNA. The Zymo-Spin™ IC features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 5 μg of DNA, and 10 μg of RNA, in ≥ 6 μL eluate. Capacity is 800 μL.
Caution: Format: DNA/RNA binding DNA binding capacity: 5 µg RNA binding capacity: 10 µg Compatibility: microcentrifuge, vacuum manifold
Customers Who Bought This Also Bought