Spécifications
- Format:Spin column
- Description:Zymo-Spin™ IC
- Size:50 pack
Spécifications
A propos de cet article
Spin columns for the purification of DNA or RNA.
- Format: DNA/RNA binding
- DNA binding capacity: 5 µg
- RNA binding capacity: 10 µg
- Compatibility: microcentrifuge, vacuum manifold
Capped version of the Zymo-Spin™ I column. The Zymo-Spin™ IC column can be used either in microcentrifuges or on vacuum manifolds for the purification of DNA and/or RNA. The Zymo-Spin™ IC features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 5 μg of DNA, and 10 μg of RNA, in ≥ 6 μL eluate. Capacity is 800 μL.