Order Entry
ContactUsLinkComponent
Microcentrifuge Tube Amber 0.5ml
Certified Microcentrifuge Tubes in Self-Standing Bags, Globe Scientific
# de catalogue 76004-092
Fournisseur:  Globe Scientific
Microcentrifuge Tube Amber 0.5ml
Certified Microcentrifuge Tubes in Self-Standing Bags, Globe Scientific
# de catalogue 76004-092
Fournisseur:  Globe Scientific

Spécifications

  • Capacity:
    0.5 ml
  • Color:
    Amber
  • Cat. no.:
    76004-092
  • Packaging:
    Bag/500

Spécifications

A propos de cet article

The polypropylene (PP) tubes are resistant to most chemical reagents and can be autoclaved at 121 °C.

  • Now available in 8 attractive colors: amber, blue, green, natural, orange, red, violet, and yellow  
  • Efficient, automated production reduces cost and assures high quality
  • Tubes are made from USP Class VI medical grade polypropylene with excellent clarity
  • Supplied in resealable, tamper-evident, self standing bags
  • Tubes are graduated with frosted marking areas and piercable caps
  • Manufactured in a class 100000 cleanroom and certified to be free from RNase, DNase, DNA, and Pyrogens

Each tube is molded with precise graduation marks for accurate measurements. These microcentrifuge tubes are offered in resealable, tamper evident self-standing bags. 

Customers Who Bought This Also Bought