Wards® Tryptic Soy Broth Powdered Media
# de catalogue 470227-480
Fournisseur: Ward's Science
Numéro CAS:
Spécifications
- Concentration:25 g/L
- Description:Tryptic Soy Broth Powder
- Size:1000 g
- Media Format:Powder
- Cat. no.:470227-480
Spécifications
A propos de cet article
High quality microbiological media at an economical price.
- SDS forms available for each media type
- Carefully blended for consistency and reliability
- Longer shelf life than prepared media
- Economical alternative
Use's Ward's® own dehydrated media for many of your routine microbiology applications.
Combine powder with water according to the mixture instructions below and sterilize for ready-to-use bacterial media.
Customers Who Bought This Also Bought