Spécifications
- Description:Water Quality Meter
- Dimensions:195 × 68 × 30 mm (7.75 × 2.5 × 1")
- Weight:250 g (8.8 oz.)
- Cat. no.:75997-568
Spécifications
A propos de cet article
This versatile meter reads pH, mV, conductivity, TDS, DO temperature and ORP by using interchangeable probes.
The user can order probes for only the parameters currently required, and add additional probes in the future. Intelligent probes electronically retain calibration information. No recalibration is required when changing probes. The display simultaneously reads temperature in user selectable °C and °F, and all measurements are automatically temperature compensated. RS232 port enables communication with a computer. Also features auto power off, hold, min/max, fold-out easel back and a tripod screw.