Pails and Lids, High-Density Polyethylene, Qorpak®
# de catalogue 75996-730
Fournisseur: Qorpak
Spécifications
- Capacity:18.9 L (5 gal.)
- Color:White
- Description:Open head pail with embossed warning label, metal handle and plastic grip
- Cat. no.:75996-730
Spécifications
A propos de cet article
Durable, sealable, high-density polyethylene pails for dispensing, storage, or waste
Rounded hand grip with metal bail. The snap-on lids feature an inner rubber sealing gasket. The Reike lid has a Reike spout with a screw cap and handy pull-out spout for easy filling and emptying. EZ Tear lid has a pull tab to secure the contents during shipping and transport.
Customers Who Bought This Also Bought