ContactUsLinkComponent
Open head Pail 5 gallon
Pails and Lids, High-Density Polyethylene, Qorpak®
# de catalogue 75996-730
Fournisseur:  Qorpak
Open head Pail 5 gallon
Pails and Lids, High-Density Polyethylene, Qorpak®
# de catalogue 75996-730
Fournisseur:  Qorpak
Spécifications
  • Capacity:
    18.9 L (5 gal.)
  • Color:
    White
  • Description:
    Open head pail with embossed warning label, metal handle and plastic grip
  • Cat. no.:
    75996-730
Spécifications
A propos de cet article
Durable, sealable, high-density polyethylene pails for dispensing, storage, or waste
Rounded hand grip with metal bail. The snap-on lids feature an inner rubber sealing gasket. The Reike lid has a Reike spout with a screw cap and handy pull-out spout for easy filling and emptying. EZ Tear lid has a pull tab to secure the contents during shipping and transport.
Customers Who Bought This Also Bought