ContactUsLinkComponent
Econo* Operating Scissors 5-1/2in
Econo™ Sterile Operating Scissors, Sklar
# de catalogue 75995-382
Fournisseur:  Sklar
Econo* Operating Scissors 5-1/2in
Econo™ Sterile Operating Scissors, Sklar
# de catalogue 75995-382
Fournisseur:  Sklar
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Material:
    Stainless Steel
  • Curvature:
    Straight
  • Cat. no.:
    75995-382
  • Length:
    140 mm (51/2")
  • Blade Type:
    Blunt/Blunt
Spécifications
A propos de cet article
Econo™ Sterile Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument.
  • Three tip configurations available
  • Able to cut tougher tissue
  • Ready-to-use
With three available tip configurations (sharp/sharp, blunt/blunt, or sharp/blunt), varying curvatures and multiple lengths, these scissors are used for general, multipurpose cutting and dissecting. Sharp Operating Scissors cut tougher tissue and are frequently used for dissecting, whereas blunt Operating Scissors are more often used for cutting materials such as sutures, tubing, or gauze. Econo™ Sterile instruments are ready-to-use, sterile, disposable, floor-grade instruments sold in bulk quantities.
Customers Who Bought This Also Bought