Spécifications
- Material:Stainless Steel
- Curvature:Straight
- Cat. no.:75995-382
- Length:140 mm (51/2")
- Blade Type:Blunt/Blunt
Spécifications
A propos de cet article
Econo™ Sterile Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument.
- Three tip configurations available
- Able to cut tougher tissue
- Ready-to-use
With three available tip configurations (sharp/sharp, blunt/blunt, or sharp/blunt), varying curvatures and multiple lengths, these scissors are used for general, multipurpose cutting and dissecting. Sharp Operating Scissors cut tougher tissue and are frequently used for dissecting, whereas blunt Operating Scissors are more often used for cutting materials such as sutures, tubing, or gauze. Econo™ Sterile instruments are ready-to-use, sterile, disposable, floor-grade instruments sold in bulk quantities.