ContactUsLinkComponent
Bulk uTIP™ Pipette Tips, Biotix
Catalogue # 75982-130
Fournisseur:  Biotix
   Waste Reducing
   Sustainable Packaging
undefined
Bulk uTIP™ Pipette Tips, Biotix
Catalogue # 75982-130
Fournisseur:  Biotix
Supplier Number:  63300082
   Waste Reducing
   Sustainable Packaging
Spécifications
  • Volume:
    1250 µl
  • Environmentally Preferable:
    Waste Reducing,
    Sustainable Packaging
  • Environmental certification:
    FSC-STD-40-004 V3-1 and FSC-C154485; Bulk tips reduce plastic waste by 95%
  • Packed:
    1000 Tips/Bag, 4 Bags/Pack
  • Cat. no.:
    75982-130
  • Tip Style:
    Low Retention, Non-Filter uTIP™ Pipette Tips
Spécifications
À propos de cet article
Biotix uTIP™ pipette tips fit the leading pipettor brands and help improve ergonomics by lowering insertion and ejection forces. This is achieved through patented technologies, including FlexFit® and Blade®. All Biotix tips contain X-Resin™ for uniform low retention properties. This results in minimal sample retention in the tip, improving the accuracy and precision of results.
  • High quality Biotix tips at a low price
  • Bulk packaging in resealable bags
  • Tips must be racked by hand
All Biotix tips are certified free of RNase, DNase, pyrogens, and PCR inhibitors.
All racks include a label with a unique bar-coded serial number, part number, and description for traceability to the individual rack.
Tips are manufactured under stringent controls in an ISO 9001 certified facility. Each lot is tested for accuracy and precision, ensuring superior quality and compatibility. All raw materials are controlled and inspected for quality. Sterile products undergo E-Beam radiation.
Frequently Bought Together