ContactUsLinkComponent
Non-Filter Tip 200ul
Bulk uTIP™ Pipette Tips, Biotix
# de catalogue 75982-126
Fournisseur:  Biotix
Non-Filter Tip 200ul
Bulk uTIP™ Pipette Tips, Biotix
# de catalogue 75982-126
Fournisseur:  Biotix
Spécifications
  • Volume:
    200 µl
  • Packed:
    1000 Tips/Bag, 10 Bags/Pack
  • Tip Style:
    Low Retention, Non-Filter uTIP™ Pipette Tips
Spécifications
A propos de cet article
Biotix uTIP™ pipette tips fit the leading pipettor brands and help improve ergonomics by lowering insertion and ejection forces. This is achieved through patented technologies, including FlexFit® and Blade®. All Biotix tips contain X-Resin™ for uniform low retention properties. This results in minimal sample retention in the tip, improving the accuracy and precision of results.
  • High quality Biotix tips at a low price
  • Bulk packaging in resealable bags
  • Tips must be racked by hand
All Biotix tips are certified free of RNase, DNase, pyrogens, and PCR inhibitors.
All racks include a label with a unique bar-coded serial number, part number, and description for traceability to the individual rack.
Tips are manufactured under stringent controls in an ISO 9001 certified facility. Each lot is tested for accuracy and precision, ensuring superior quality and compatibility. All raw materials are controlled and inspected for quality. Sterile products undergo E-Beam radiation.
Customers Who Bought This Also Bought