Hose Tubing Connectors, Bürkle
# de catalogue 75876-094
Fournisseur: BURKLE INC
Spécifications
- Tubing I.D.:7–10 mm (0.28–0.39") Diameter
- Cat. no.:75876-094
- Supplier No.:8700-0810
Spécifications
A propos de cet article
Conical, straight hose connectors for various hose sizes.
- Polypropylene construction
- Sterilizable
Packaging: 10 connectors per package.
Customers Who Bought This Also Bought