Hose Tubing Connectors, Bürkle
Catalog # 75876-090
Supplier: BURKLE INC
Specifications
- Tubing I.D.:3 - 5 mm (0.12 - 0.20") diameter
- Cat. no.:75876-090
- Supplier No.:8700-0406
Specifications
About this item
Conical, straight hose connectors for various hose sizes.
- Polypropylene construction
- Sterilizable
Packaging: 10 connectors per package.
Customers Who Bought This Also Bought