Spécifications
- Description:2GP Urinalysis Strips
- Cat. no.:95064-055
- Parameters Tested:Protein, Glucose
- Supplier No.:DTG-2GP
Spécifications
A propos de cet article
Test strips for urinalysis testing of various parameters.
- Consistent, highly accurate and sensitive
- Competitively priced
- Can be read visually or by using the Clarity 120C Urine Analyzer
- 16 to 18 month dating
- 90 day open vial stability
- Room temperature storage
- Read results between 60 to 20 seconds
- Color chart made in Germany
- CAP/API values listed
Strips detect the following 10 parameters: Leukocytes/Nitrite/Urobilinogen/Protein/Ph/Blood/Specific Gravity/Ketone/Bilirubin/Glucose in urine samples.
Ideal for POL, Physician offices, Urgent Care Locations, OB-GYN offices, and Hospitals.
Tests strips have an 18-month shelf life.