ContactUsLinkComponent
STRIP UROCHECK URINALYSIS 2GP CLARITY
Clarity Urocheck Urinalysis Strips, Clarity Diagnostics
# de catalogue 95064-055
Fournisseur:  Clarity Diagnostics
Numéro CAS:  
STRIP UROCHECK URINALYSIS 2GP CLARITY
Clarity Urocheck Urinalysis Strips, Clarity Diagnostics
# de catalogue 95064-055
Fournisseur:  Clarity Diagnostics
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    2GP Urinalysis Strips
  • Cat. no.:
    95064-055
  • Parameters Tested:
    Protein, Glucose
  • Supplier No.:
    DTG-2GP
Spécifications
A propos de cet article
Test strips for urinalysis testing of various parameters.
  • Consistent, highly accurate and sensitive
  • Competitively priced
  • Can be read visually or by using the Clarity 120C Urine Analyzer
  • 16 to 18 month dating
  • 90 day open vial stability
  • Room temperature storage
  • Read results between 60 to 20 seconds
  • Color chart made in Germany
  • CAP/API values listed
Strips detect the following 10 parameters: Leukocytes/Nitrite/Urobilinogen/Protein/Ph/Blood/Specific Gravity/Ketone/Bilirubin/Glucose in urine samples.
Ideal for POL, Physician offices, Urgent Care Locations, OB-GYN offices, and Hospitals.
Tests strips have an 18-month shelf life.
Certifications: CLIA waived. FDA approved.
Packaging: 100 strips per bottle.
Customers Who Bought This Also Bought