ContactUsLinkComponent
VWR* Microplate 96 Square Deep Well, Material: Polypropylene, DNA/RNA Free, Sterile, Capacity: 2Ml
VWR® 96-Well Deep Well Microplates, Extractable-Free
# de catalogue 75870-796
Fournisseur:  VWR International
VWR* Microplate 96 Square Deep Well, Material: Polypropylene, DNA/RNA Free, Sterile, Capacity: 2Ml
VWR® 96-Well Deep Well Microplates, Extractable-Free
# de catalogue 75870-796
Fournisseur:  VWR International
Spécifications
  • Sterility:
    Sterile
  • Well Volume:
    2 mL
  • Description:
    96-Well Deep Well Microplates
  • Cat. no.:
    75870-796
  • Well Shape:
    Square
  • Well Bottom Shape:
    Pyramid
Spécifications
A propos de cet article
A range of high-quality, dependable deep-well blocks for storage and collection, LC-MS, SPE, and mixing.
  • Guaranteed extractable-free
  • Gamma irradiated
  • Ready-to-use
  • For assured assays
  • DNase-free and RNase-free
These microplates are made of extractable-free polypropylene that will not leach unwanted compounds from the polymer.
Plates can be sealed with friction or heat seals to prevent evaporation, condensation, and contamination. These plates are ideal for sample storage and transport. Blocks are rigid and robust, and they can be easily stacked, handled, manipulated, heat sealed, deep frozen, autoclaved, and put in a genogrinder.
This protects assays from false results due to contamination. No sterilization is required. These plates save time and reduce costs by eliminating the cleaning process and sterility validation. These plates conform to internationally recognized standards and fit all standard laboratory equipment without adjustment.
Certifications: Made in Class 10000 cleanroom. Standard ANSI/SLAS footprint and dimensions.
Packaging: Sterile Microplates are gamma irradiated and packaged in bags of 5.
Customers Who Bought This Also Bought