Spécifications
- Sterility:Sterile
- Well Volume:2 mL
- Description:96-Well Deep Well Microplates
- Cat. no.:75870-796
- Well Shape:Square
- Well Bottom Shape:Pyramid
Spécifications
A propos de cet article
A range of high-quality, dependable deep-well blocks for storage and collection, LC-MS, SPE, and mixing.
- Guaranteed extractable-free
- Gamma irradiated
- Ready-to-use
- For assured assays
- DNase-free and RNase-free
These microplates are made of extractable-free polypropylene that will not leach unwanted compounds from the polymer.
Plates can be sealed with friction or heat seals to prevent evaporation, condensation, and contamination. These plates are ideal for sample storage and transport. Blocks are rigid and robust, and they can be easily stacked, handled, manipulated, heat sealed, deep frozen, autoclaved, and put in a genogrinder.
This protects assays from false results due to contamination. No sterilization is required. These plates save time and reduce costs by eliminating the cleaning process and sterility validation. These plates conform to internationally recognized standards and fit all standard laboratory equipment without adjustment.