Order Entry
ContactUsLinkComponent
Measure-Matic* Dispenser
Measure-Matic™ Measuring Dispenser, Electron Microscopy Sciences
# de catalogue 103302-510
Measure-Matic* Dispenser
Measure-Matic™ Measuring Dispenser, Electron Microscopy Sciences
# de catalogue 103302-510
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Description:
    Dispenser
  • Cat. no.:
    103302-510
  • Supplier No.:
    64299-08

Spécifications

A propos de cet article

Makes repeated measuring and pouring easy!

  • Adjust the dispensing cup to the desired volume, then squeeze the 500 mL (16 oz) reservoir bottle
  • The desired volume of liquid will flow through the tube and into the cup
  • Excess liquid will flow back into the reservoir bottle
  • Cup and closure are made of rugged polypropylene
  • Reservoir bottle is made of squeezable-density polyethylene

Adjustable for volumes from 5 mL to 25 mL. Just squeeze and check the fluid level. Dust cover included.

Delivery: Dust cover included.
Customers Who Bought This Also Bought