ContactUsLinkComponent
Plastic Dropper, With Cap and Bulb, For 60ml dropper bottle
Accessories for Dropping Bottles, Amber, Kimble Chase
# de catalogue 75855-878
Accessories for Dropping Bottles, Amber, Kimble Chase
# de catalogue 75855-878
Spécifications
  • Description:
    Plastic Dropper with Rubber Nipple, 60 mL
  • Cat. no.:
    75855-878
  • Supplier No.:
    15040D-6003