Spécifications
- Sterility:Sterile
- Volume:1 ml
- Cat. no.:89215-217
- For Use With:Human, Laboratory and Veterinary Use
- Type:Luer Slip Tip, Tuberculin, Low Dead Space Plunger
- Supplier No.:8300014579
Spécifications
A propos de cet article
HENKE-JECT® syringes are a high quality disposable rubber-piston syringe offering substantial savings.
- ETO gas sterilized
- Latex and pyrogen free
A lubricated synthetic rubber piston allows for a smooth draw while a positive stop helps prevent accidental pullout. Bold graduations are easily read and resist wipe off.