Cryokit 550, 22" Cryogenic Gloves, Mapa Professional
# de catalogue 75841-230
Fournisseur: MAPA
Spécifications
- Ambidextrous/Hand-specific:Hand-specific
- Glove Color:Blue,Black
- Glove size:8
- Length:22"
- Disposable / reusable:Reusable
- Material:Polyester
- Cuff style:Gauntlet
- Liner material:Polyolefin
- Finish:Elastic Laminated Blue Fabric
- Cat. no.:75841-230
- Supplier No.:CRYKIT550558
Spécifications
A propos de cet article
These gloves are designed for applications handling liquid nitrogen and other cryogenic gases, to protect from cold contact and prevent burns from liquid gas leakage.
- Multilayer fleece lining
- Ventilation technology
- Waterproof
Gloves remain ventilated due to the inner polyolefin porelle membrane which allows for air to flow and keeps hands dry.
A special elastic laminated blue fabric provides dexterity and comfort in the glove and cuff areas.
Certifications: CE Cat. III 0498 marked. Cold Hazards EN511: 231. Mechanical Hazards EN388: 2422.
Customers Who Bought This Also Bought