ContactUsLinkComponent
Retro-Reflective Sensor PBT/PC enclosure with NPN output
Photo Reflective Sensor, Nidec Shimpo
# de catalogue 75840-664
Fournisseur:  Nidec Shimpo America
Retro-Reflective Sensor PBT/PC enclosure with NPN output
Photo Reflective Sensor, Nidec Shimpo
# de catalogue 75840-664
Fournisseur:  Nidec Shimpo America
Spécifications
  • Description:
    Photo reflective sensor; NPN open collector to 1000 Hz, M18x1 connection
  • Cat. no.:
    75840-664
  • Range:
    0.05 - 7 m (2 - 23")
  • Supplier No.:
    MCS-655P
Spécifications
A propos de cet article
Photo Sensors can be adjusted to detect differences in material colors.
  • LED indication for ease of set-up
Model LS-S50ML is a laser sensor that can be taught to recognize difference in colors
Model MCS-655P is a photo-reflective NPN open collector photo sensor with Light/Dark activated output that operates up to 1000 Hz. This model comes in a robust metal housing with a connection cable, LED status indicator, sensitivity control, and mounting nuts.
Model MCS-655M is a photo-reflective NPN open collector photo sensor with Light/Dark activated output that operates up to 1000 Hz. This model comes in a robust metal housing with a connection cable, LED status indicator, sensitivity control, and mounting nuts.
It is a class 1 red laser with switching frequency of 1.5kHz. It is contained in metal housing with 6' of cable, and can be utilized with stroboscopes, counters, and panel tachometers. It includes a trimmer potentiometer located on the side for minimum and optimum sensitivity adjustments. This sensor can be used in normally closed or normally open operation, NPN type of output, with a yellow LED for signal indication and a green LED for power.
Customers Who Bought This Also Bought