ContactUsLinkComponent
Proximity Sensor three wire DC
Proximity Sensors, Nidec Shimpo
# de catalogue 75840-644
Fournisseur:  Nidec Shimpo America
Proximity Sensor three wire DC
Proximity Sensors, Nidec Shimpo
# de catalogue 75840-644
Fournisseur:  Nidec Shimpo America
Spécifications
  • Description:
    Proximity Sensor; NPN (NO) Sinking Output, 1.5 kHz frequency
  • Cat. no.:
    75840-644
  • Range:
    2 mm (0.08")
  • Supplier No.:
    BI2-S12
Spécifications
A propos de cet article
Ideal for gear speed measurement.
  • Rugged Proximity Sensors to measure position, movement, or rotational speeds
  • Economical solution for detecting metal in front of a sensing surface
  • Metal objects adjacent to or behind will not normally be detected
Model DJ2 features a pre-wired 17' cable assembly, zero-sensing capability, and small package size
Model B12-S12 is a three-wire DC normally open NPN (sinking) type proximity switch which features a short circuit and overload protection plus LED output indicator.
Model SE-G is a solid state proximity switch especially designed for gear sensing. This sensor will provide a square wave signal output and indicate detection via LED indicator on rear of unit. Maximum switching frequency is 8 kHz with a maximum sensing distance of 1.0mm. Gear pitch must be between 25.4 (min.) and 16.93 (max.).
Model MCS-3109 is a complimentary (NPN-NO and NPN-NC) proximity sensor normally used in high-vibration areas and where a greater sensing distance is required.
Customers Who Bought This Also Bought