ContactUsLinkComponent
Pure Preserved Pig Kidney Plain V
Ward's® Pure Preserved™ Pig Kidney
# de catalogue 470223-854
Fournisseur:  Avantor
Ward's® Pure Preserved™ Pig Kidney
# de catalogue 470223-854
Fournisseur:  Avantor
Spécifications
  • Injection Type:
    Uninjected
  • Cat. no.:
    470223-854
  • Packaging:
    Vacuum Pack of 10
  • Supplier No.:
    WARD470223-854
Spécifications
A propos de cet article
Ward’s® PURE Preserved Specimens - the best dissection experience from start to finish.
  • Safest specimens
  • Low formalin odor
  • Best classroom experience
Ward's® PURE Preserved Specimens offer the best classroom experience because PURE Specimens are safe. Students stay focused on learning and not on smells. Experience quality with true-to-life preservation, combined with artfully illustrated dissection guides, free with every purchase, to maximize value, lesson time and student engagement.
We guarantee you’ve never experienced dissection like this before. Go Pure.
Pig kidneys may contain a small cut as part of a USDA inspection procedure. This will not compromise specimen quality.
Packaging: Our preserved pig kidneys are shipped vacuum packed or in pails.
Customers Who Bought This Also Bought