Order Entry
ContactUsLinkComponent
COMPOSITE 5 K RGT HYDRANAL F/KF TITRN 1L
HYDRANAL® Karl Fischer Reagents, Honeywell Research Chemicals
# de catalogue BJ34816-1L
Numéro CAS:  
HYDRANAL® Karl Fischer Reagents, Honeywell Research Chemicals
# de catalogue BJ34816-1L
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Concentration:
    5 mg/ml
  • Packaging:
    Polycoated bottle
  • Description:
    HYDRANAL™-Composite 5 K, Reagent for volumetric one-component Karl Fischer titration in aldehydes and ketones (methanol free)
  • Size:
    6x 1 L
  • Cat. no.:
    BJ34816-1L
  • Supplier No.:
    34816-6X1L

Spécifications

A propos de cet article

Original and Innovative Karl Fischer Reagents to analyze or measure water content

Our premium Hydranal™ product range for pyridine-free and ethanol-based water determination by Karl Fischer titration (KF titration) is comprised of one- and two-component reagents for volumetric Karl Fischer titrations. Also available are water standards for titer standardization, pH buffering reagents, special reagents for aldehydes, ketones, and other solubilizing agents.

Customers Who Bought This Also Bought