ContactUsLinkComponent
Wide mouth bottle 1/2gallon
Bottle, Wide Mouth, PTFE, C&G Containers
# de catalogue 75839-274
Fournisseur:  C&G Containers
Bottle, Wide Mouth, PTFE, C&G Containers
# de catalogue 75839-274
Fournisseur:  C&G Containers
Spécifications
  • Cap Size:
    83 - 400
  • Capacity:
    1.89 L (½ gal.)
  • Color:
    Clear
  • Description:
    Bottes with A1 wash protocol, wide mouth with solid cap
  • Cap Color:
    White
  • Supplier No.:
    LFW064100000