Methanol ≥99.9%, HiPerSolv CHROMANORM® for HPLC-MS, suitable for LC-MS, UPLC, UHPLC, Ultra HPLC, VWR Chemicals BDH®
# de catalogue BDH85800.400
Fournisseur: VWR International
Numéro CAS: 67-56-1
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Glass bottle
- Size:4 L
- Cat. no.:BDH85800.400
- Supplier No.:BDH85800.400
Spécifications
Résultats des tests de spécification
- Assay (GC):99.9% Min.
- Appearance:Clear colourless liquid
- Carbonyl compounds:Passes test
- Solubility in water:Passes test
- Substances darkened by sulphuric acid:Passes test
- Substances reducing permanganate:Passes test
- Acidity:0.0002 meq/g Max.
- Alkalinity:0.0002 meq/g Max.
- Boiling point:64 - 65 °C
- Colouration:0 APHA Max.
- Density (20/20):0.791 - 0.793
- Evaporation residue:1 ppm Max.
- Water:0.02 % Max.
- Al (Aluminium):20 ppb Max
- Ag (Silver):100 ppb Max
- As (Arsenic):20 ppb Max
- Ba (Barium):100 ppb Max
- Ca (Calcium):200 ppb Max
- Cd (Cadmium):20 ppb Max
- Co (Cobalt):20 ppb Max
- Cr (Chromium):20 ppb Max
- Cu (Copper):10 ppb Max
- Fe (Iron):50 ppb Max
- K (Potassium):50 ppb Max
- Mg (Magnesium):50 ppb Max
- Mn (Manganese):100 ppb Max
- Mo (Molybdenum):20 ppb Max
- Na (Sodium):200 ppb Max
- Ni (Nickel):100 ppb Max
- Pb (Lead):20 ppb Max
- V (Vanadium):20 ppb Max
- Zn (Zinc):100 ppb Max
- Gradient (220 nm):3 mAU Max
- Gradient (235 nm):2 mAU Max
- Gradient (254 nm):1 mAU Max
- Transmittance (210 nm):45% Min
- Transmittance (220 nm):65% Min
- Transmittance (225 nm):70% Min
- Transmittance (230 nm):85% Min
- Transmittance (235 nm):90% Min
- Transmittance (240 nm):95% Min
- Transmittance (250 nm):95% Min
- Transmittance (260 nm):98% Min
- Transmittance (280-400 nm):98% Min
- Absorbance (210 nm):0.347 Max
- Absorbance (220 nm):0.188 Max
- Absorbance (225 nm):0.155 Max
- Absorbance (230 nm):0.071 Max
- Absorbance (235 nm):0.046 Max
- Absorbance (240 nm):0.023 Max
- Absorbance (250 nm):0.023 Max
- Absorbance (260 nm):0.009 Max
- Absorbance (280-400 nm):0.009 Max
- Fluorescence (as quinine) (254 nm):1 ppb Max
- Fluorescence (as quinine) (365 nm):0.5 ppb Max
- MS-ESI+ (as Reserpine):50 ppb Max
- MS-APCI+ (as Reserpine):50 ppb Max
- MS-ESI- (as 4-Nitrophenol):20 ppb Max
- MS-APCI- (as 4-Nitrophenol):20 ppb Max
- Conforms Ph.Eur. R1 1053201:Passes test
- Conforms Ph.Eur. R2 1053202:Passes test
- Conforms to ACS:Passes test
- Filtered at 0.2 μm:Confirmed
- Methanol ≥99.9%, HiPerSolv CHROMANORM® for HPLC-MS, suitable for LC-MS, UPLC, UHPLC, Ultra HPLC, VWR Chemicals BDH®:
Résultats des tests de spécification
A propos de cet article
Synonymes: Hydroxy methane,Carbinol,Wood alcohol
Formule:CH₃OH
Numéro MDL:MFCD00004595
Numéro CAS:67-56-1
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Pt ébul.:|Point d'ébullition:
:Pt ébul.: 64,6 °C (1013 hPa)
:Pt Fusion:|Point de fusion:
:Pt Fusion: -98 °C
:Pt Eclair:|Point eclaire:
:Pt Eclair: 11 °C (coupe fermée)
:M.W. 32,04 g/mol
:-98
:64,6
ONU:1230
:11
:Length
:Width
:Height
:Diameter
:Thickness
:99,9
:Boiling Pt:
:Boiling Pt: 64.6 °C (1013 hPa)
:Melting Pt:
:Melting Pt: -98 °C
:Flash Pt:
:Flash Pt: 11 °C (closed cup)
:M.W. 32.04 g/mol
:64.6
:3
:II
Customers Who Bought This Also Bought