GSBP-inowax glycols 30Mx0.25MMx0.25UM
# de catalogue 75839-214
Fournisseur: General Separation Technologies, Inc.
GSBP-inowax glycols 30Mx0.25MMx0.25UM
# de catalogue 75839-214
Fournisseur: General Separation Technologies, Inc.
Customers Who Bought This Also Bought
VWR® Screw Caps for 9 mm Screw-Thread Vials, Wide Opening
Fournisseur: VWR International
$57.66 to $576.72
SureSTART™ Screw Caps, 9 mm, Level 3 High Performance Applications
Fournisseur: Thermo Scientific
$79.21 to $82.14
VWR® 10 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
$127.69
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
$137.63
SureSTART™ Crimp Caps, 20 mm, Level 2 High-throughput Applications, Thermo Scientific
Fournisseur: Thermo Scientific
$180.01 to $182.41
WHEATON® MicroLiter 9 mm Screw-Thread Vials, Component Kits, Unassembled, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$167.84
Screw Caps with Partial Slit Septa for 12×32 mm Vials, Microsolv Technology Corp
Fournisseur: MicroSolv Technology Corp
$119.01
SureSTART™ Glass Screw Top 0.3 ml Microvials for <2 ml Samples, Level 3 High Performance Applications, Thermo Scientific
Fournisseur: Thermo Scientific
$182.38 to $196.03
SureSTART™ Glass Screw Top Vials, 2 ml, Level 3 High Performance Applications, Thermo Scientific
Fournisseur: Thermo Scientific
$43.20 to $44.79