GSBP-AL2O3 M 60Mx0.53MMx15.00UM
# de catalogue 75839-136
Fournisseur: General Separation Technologies, Inc.
GSBP-AL2O3 M 60Mx0.53MMx15.00UM
# de catalogue 75839-136
Fournisseur: General Separation Technologies, Inc.
Customers Who Bought This Also Bought
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
121,85 $ to 1 194,50 $
VWR® Screw Caps for 9 mm Screw-Thread Vials, Wide Opening
Fournisseur: VWR International
86,08 $ to 717,60 $
SureSTART™ Screw Caps, 9 mm, Level 3 High Performance Applications
Fournisseur: Thermo Scientific
79,21 $ to 82,14 $
SureSTART™ Septa for 20 mm Stoppers, Level 2 High-throughput Applications, Thermo Scientific
Fournisseur: Thermo Scientific
131,41 $ to 136,31 $
SureSTART™ Screw Caps for Glass Screw Top Vials, 9 mm, Level 1 Everyday Analysis, Thermo Scientific
Fournisseur: Thermo Scientific
363,91 $ to 363,91 $