Porapak N 80/100 1Mx1/8IN-ODx2MM-ID
# de catalogue 75839-076
Fournisseur: General Separation Technologies, Inc.
Porapak N 80/100 1Mx1/8IN-ODx2MM-ID
# de catalogue 75839-076
Fournisseur: General Separation Technologies, Inc.
Customers Who Bought This Also Bought
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
$50.14 to $501.54
SureSTART™ Caps for 2 ml Glass Snap Top Vials, Level 2 High-Throughput Applications, Thermo Scientific
Fournisseur: Thermo Scientific
$45.12 to $48.54
WHEATON® MicroLiter Crimp-Top Vials, 11 mm, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$68.56