Hayesep Q 80/100 10FTx1/8IN-ODx2MM-ID
# de catalogue 75839-074
Fournisseur: General Separation Technologies, Inc.
Hayesep Q 80/100 10FTx1/8IN-ODx2MM-ID
# de catalogue 75839-074
Fournisseur: General Separation Technologies, Inc.
Customers Who Bought This Also Bought
WHEATON® MicroLiter Septa for 20 mm Crimp Seals, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
343,64 $
WHEATON® ABC Vial™ Large-Opening Screw-Thread Vials, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
153,23 $
MicroSolv™ Pharma Grade Vials, Caps and Convenience Kits
Fournisseur: MicroSolv Technology Corp
146,67 $
WHEATON® MicroLiter Screw-Thread Vials, Component Kits, Assembled, 9 mm, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
127,72 $
WHEATON® MicroLiter Snap-Top Vials, 11 mm, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
231,64 $
SureSTART™ Screw Caps, 9 mm, Level 3 High Performance Applications
Fournisseur: Thermo Scientific
79,21 $ to 82,14 $