GSBP-VMS 30Mx0.32MMx1.80UM
# de catalogue 75839-052
Fournisseur: General Separation Technologies, Inc.
GSBP-VMS 30Mx0.32MMx1.80UM
# de catalogue 75839-052
Fournisseur: General Separation Technologies, Inc.
Customers Who Bought This Also Bought
VWR® Snap Caps for 11 mm Snap Ring Vials, Wide Opening
Fournisseur: VWR International
$73.75 to $568.62
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
$50.14 to $501.54
WHEATON® ABC Vial™ Large-Opening Screw-Thread Vials, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$153.23
VWR® 11 mm Crimp Top Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
$291.56
SureSTART™ Glass Screw Top Vials, 2 ml, Level 3 High Performance Applications, Thermo Scientific
Fournisseur: Thermo Scientific
$43.20 to $44.79
WHEATON® E-Z Vial® Large-Opening Crimp-Top Vials, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$125.61