Order Entry
Canada
ContactUsLinkComponent
Polypropylene Molded Graduated Cylinders, Globe Scientific
Catalogue # 75838-810
Fournisseur:  Globe Scientific
undefined
Polypropylene Molded Graduated Cylinders, Globe Scientific
Catalogue # 75838-810
Fournisseur:  Globe Scientific
Supplier Number:  601078

Spécifications

  • Capacity:
    50 ml
  • Cat. no.:
    75838-810
  • Supplier No.:
    601078

Spécifications

À propos de cet article

These superior quality graduated cylinders are ideal for the safe and accurate measurement of all types of liquids.

  • Wide opening and tapered pour spout
  • Break-resistent
  • Can withstand a continuous working environment up to 100°C

Cylinders feature a wide rim and a tapered pour spout for easy filling and pouring. The non-wetting interior surface eliminates concave menisci, providing more accurate measurements. The wide pentagonal-shaped base provides stability and makes the cylinder roll-resistant. Heavy-duty plastic construction withstands repeated autoclaving. Switching from glass cylinders to Globe’s plastic equivalents will greatly reduce breakage and the potential for injury.

Certifications: Conforms to ISO 6706-1981.
Frequently Bought Together