Pitchers with Molded Graduations, Globe Scientific
# de catalogue 75838-762
Fournisseur: Globe Scientific
Spécifications
- Capacity:5000 ml
- Cat. no.:75838-762
- Supplier No.:601160-1
Spécifications
A propos de cet article
These pitchers are ideal for all general laboratory procedures.
- Molded in handle with thumb grip
- Special no-drip pour spout
- Graduated in milliliters (ml)
In one pitcher you can mix, measure and prepare solutions. The pitchers have a no-drip pour spout and a molded-in handle with thumb grip. They are made from heavy-duty plastic and withstands repeated autoclaving. Switching from glass beakers to Globe’s plastic equivalents will greatly reduce breakage and the potential for injury.
Customers Who Bought This Also Bought