Pitchers with Molded Graduations, Globe Scientific
Catalog # 75838-758
Supplier: Globe Scientific
Specifications
- Capacity:2000 ml
- Cat. no.:75838-758
- Supplier No.:601158-1
Specifications
About this item
These pitchers are ideal for all general laboratory procedures.
- Molded in handle with thumb grip
- Special no-drip pour spout
- Graduated in milliliters (ml)
In one pitcher you can mix, measure and prepare solutions. The pitchers have a no-drip pour spout and a molded-in handle with thumb grip. They are made from heavy-duty plastic and withstands repeated autoclaving. Switching from glass beakers to Globe’s plastic equivalents will greatly reduce breakage and the potential for injury.
Customers Who Bought This Also Bought