Sapphire PCR Tubes and Strips, Polypropylene, Greiner Bio-One
Catalog # 75786-257
Supplier: Greiner Bio-One
Specifications
- Capacity:0.2 ml
- Color:Clear
- Description:PCR Strips, Standard, Clear, with Individual Attached Optical Caps for Real Time PCR
- Supplier No.:608281
Specifications
About this item
Sapphire PCR Tubes and strips feature attached or unattached caps in either domed or flat designs, and frosted caps are available for easy labeling.
- Manufactured out of medical-grade polypropylene without interfering additives
- Thin-walled for efficient heat transfer during amplification
- Options for real-time/quantitative PCR
- Comprehensive thermal cycler compatibility
- Packaging labeled with product symbols for easier working and warehousing
Maximum volume capacity ranges between models. Tube strips are available in formats of standard or low profile.
Free of detectable DNase, RNase, human DNA, endotoxin and PCR inhibitors.
Frequently Bought Together