VWR® Classic Syringe Tips, Non sterile
Catalog # 75838-210
Supplier: VWR International
Specifications
- Volume:50 ml
- Cat. no.:75838-210
- Sterile:-
Specifications
About this item
Classic Syringe Tips provide reliable and repeatable results, and they are compatible with a variety of pipettors.
- Ideal for use with viscous, infectious, and volatile liquids
- Compatible with a variety of pipettors
Ripette®, Ripette®pro, Ripette®genX, eLineTM-Dispenser, Minilab 100, Minilab 101, Minilab 201, Distriman®, StepMate, EasyStep, EasyStep electronic, RepeatOne®, Handrop.
These syringe tips are not compatible with: HandyStep®, Repetman®.
Ordering information: The 25 and 50 ml tips require an adapter (needs to be ordered separately).
Customers Who Bought This Also Bought