Spécifications
- Accuracy:0.01% of FS
- Range:60-120,000 FPM
- Resolution:60-12,000 = 0.1 FPM, 12,000-120,000 = 1FPM
- Description:Super Bright LED Portable Stroboscope
- Cat. no.:75836-646
- Supplier No.:DT-326B
Spécifications
A propos de cet article
The DT-326B’s LED array provides a bright, stable strobe light over a wide measurement range with a lifetime far exceeding xenon lit stroboscopes.
- Wide Flash Range 60-120,000 FPM
- Long-life Black Light LED operation
- Adjustable Flash Pulse duration achieves sharper images for detailed inspection
- Input sensor trigger function allows process machinery to control flash rate
The DT-326B contains a rechargeable lithium battery, a single charge can last up to a full 12 hours of operation. The DT-326B is designed for speed and frequency measurements in motion and vibration analysis applications. It is ideal for predictive and preventive maintenance applications such as: motors, shafts, roto-gravure printing, extruders, blow molding, wire lettering and striping, engraving, pulse jets, water jets, fans, cams, gear teeth, belt inspections, fuel injectors, vibration analysis, audio speaker analysis, spindle spinning, cutting blade timing and sharpness, plus many more in the printing, packaging, textile, automotive, cable, mining, steel, chemical, optical, medical and shipbuilding industries.