Superwhite Coverglass, Springside Scientific
# de catalogue 75836-078
Fournisseur: SPRINGSIDE SCIENTIFIC, LLC.
Spécifications
- Numbering:#1
- Description:Superwhite cover glasses, rectangular
- L×W:50×24 mm
- Cat. no.:75836-078
- Supplier No.:HCW122450
Spécifications
A propos de cet article
Microscope coverslips are made of pure, super white glass which prevents auto-fluorescence
They work well in all automated coverslippers with no sticking or breaking. Varied sizes are available.
Ordering information: Each plastic box contains 1oz. Each case includes 10 boxes, and is vaccum sealed.
Customers Who Bought This Also Bought