Spécifications
- Concentration:3 µg/ml each in acetonitrile*
- Description:CARB 1004 Aldehyde/Ketone-DNPH Calibration Standard
- Size:1 ml/Ampule*
- Cat. no.:75835-152
- Supplier No.:33093
Spécifications
A propos de cet article
The CARB 1004 aldehyde/ketone-DNPH calibration standard consists of 13 components, they are acetaldehyde-2,4-DNPH, acetone-2,4-DNPH, acrolein-2,4-DNPH, benzaldehyde-2,4-DNPH, n-butyraldehyde-2,4-DNPH, crotonaldehyde-2,4-DNPH, formaldehyde-2,4-DNPH, hexaldehyde-2,4-DNPH, methacrolein-2,4-DNPH, methyl ethyl ketone-2,4-DNPH, propionaldehyde-2,4-DNPH, m-tolualdehyde-2,4-DNPH, valeraldehyde-2,4-DNPH.