Order Entry
ContactUsLinkComponent
Standard Formaldehyde-DNPH Mix 1 mL/ampul
Formaldehyde-DNPH Mix, Restek
# de catalogue 75834-810
Fournisseur:  Restek
Numéro CAS:  
Formaldehyde-DNPH Mix, Restek
# de catalogue 75834-810
Fournisseur:  Restek
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Concentration:
    500 µg/ml in acetonitrile*
  • CAS No.:
    1081-15-8
  • Description:
    Formaldehyde-DNPH
  • Size:
    1 ml/Ampule
  • Formula:
    C₇H₆N₄O₄
  • Cat. no.:
    75834-810
  • Supplier No.:
    31837

Spécifications

A propos de cet article

This mix with acetonitrile solvent suitable for HPLC analysis.

  • High concentration

Sampling for airborne carbonyl compounds in EPA Method TO-11A and other methods is based on a reaction between the target compounds and 2,4-dinitrophenylhydrazine (DNPH), coated on a silica gel adsorbent. Meet the needs of investigators analyzing only for formaldehyde.

Customers Who Bought This Also Bought