Order Entry
ContactUsLinkComponent
Standard PAH Calibration Mix B 610, 1 mL/ampule
610 PAH Calibration Mix B (16 Components), Restek
# de catalogue 75834-700
Fournisseur:  Restek
Numéro CAS:  
610 PAH Calibration Mix B (16 Components), Restek
# de catalogue 75834-700
Fournisseur:  Restek
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Concentration:
    In methylene chloride:methanol (1:1)
  • Description:
    610 PAH Calibration Mix B
  • Size:
    1 ml/Ampule
  • Cat. no.:
    75834-700
  • Supplier No.:
    31455

Spécifications

A propos de cet article

This 610 PAH calibration mix B idela for LC-UV detection.

It consists of 16 components: Acenaphthene (1000 μg/ml), Acenaphthylene (2000 μg/ml), Anthracene(100 μg/ml), Benz(a)anthracene (100 μg/ml), Benzo(a)pyrene (100 μg/ml), Benzo(b)fluoranthene (200 μg/ml), Benzo(k)fluoranthene (100 μg/ml), Benzo(ghi)perylene (200 μg/ml), Chrysene (100 μg/ml), Dibenz(a,h)anthracene (200 μg/ml), Fluoranthene (200 μg/ml), Fluorene (200 μg/ml), Indeno(1,2,3-cd)pyrene (100 μg/ml), Naphthalene (1000 μg/ml), Phenanthrene (100 μg/ml) and Pyrene (100 μg/ml).

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Customers Who Bought This Also Bought