Order Entry
ContactUsLinkComponent
DHA PiONA Standard, 0.15 mL in Autosampler Vial
DHA PiONA Standard (133 Components), Restek
# de catalogue 75834-588
Fournisseur:  Restek
Numéro CAS:  
DHA PiONA Standard (133 Components), Restek
# de catalogue 75834-588
Fournisseur:  Restek
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Concentration:
    Neat
  • Description:
    DHA PiONA Standard
  • Size:
    0.15 ml in autosampler vial
  • Cat. no.:
    75834-588
  • Supplier No.:
    30730

Spécifications

A propos de cet article

This standard is produced for refineries performing Detailed Hydrocarbon Analysis (DHA) of crude oil feedstocks and fuels.

It contains 133 components.

It helps with calibrating complex hydrocarbon analyses and provides the greatest number of gravimetrically determined values for quantitative calibration.

Certifications: ASTM Methods D6729, D6730, and D6733 (Determination of Individual Components in Spark Ignition Engine Fuels)
Ordering information: Quantity discounts not available.
Customers Who Bought This Also Bought