Spécifications
- Packaging:5 x 50 disks
- Description:Vancomycin, Va-30
- Cat. no.:75832-722
- Supplier No.:Z9245
Spécifications
A propos de cet article
HardyDisk™ AST’s are impregnated paper disks used for antimicrobial susceptibility testing (AST), also known as disk diffusion or Kirby-Bauer testing
Fully compatible with the BBL single or multiple disk dispensers.