ContactUsLinkComponent
Scrub Pant Elastic Waist with Pocket 6XL
VWR® Disposable Scrub Pants
# de catalogue 75832-250
Fournisseur:  VWR International
Scrub Pant Elastic Waist with Pocket 6XL
VWR® Disposable Scrub Pants
# de catalogue 75832-250
Fournisseur:  VWR International
Spécifications
  • Clothing Size:
    6XL
  • Color:
    Blue
  • Cat. no.:
    75832-250
  • Supplier No.:
    7388
Spécifications
A propos de cet article
VWR® Disposable Scrub Pants are an ideal option for single-use needs.
  • Elastic Waist
  • Single Use
  • Hip Pockets Available
  • USA Made
  • Perfect with VWR® Disposable Scrub Shirts
These garments feature high-quality sewn-seam construction and are made from SMS nonwoven fabric for comfort.
The versatile pants help meet PPE requirements and provide comfort at the same time.
Packaging: These blue scrubs pants are packaged 30 per case within a double bag-lined carton.
Customers Who Bought This Also Bought