VWR® Disposable Scrub Pants
# de catalogue 75832-246
Fournisseur: VWR International
Spécifications
- Clothing Size:4XL
- Color:Blue
- Supplier No.:7386
Spécifications
A propos de cet article
VWR® Disposable Scrub Pants are an ideal option for single-use needs.
- Elastic Waist
- Single Use
- Hip Pockets Available
- USA Made
- Perfect with VWR® Disposable Scrub Shirts
These garments feature high-quality sewn-seam construction and are made from SMS nonwoven fabric for comfort.
The versatile pants help meet PPE requirements and provide comfort at the same time.
Packaging: These blue scrubs pants are packaged 30 per case within a double bag-lined carton.
Customers Who Bought This Also Bought